2-Methyl-1,3-dithiolane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23372-06
B23372-14
AAB23372-06EA
61.3
USD
AAB23372-06
AAB23372-14
2-Methyl-1,3-dithiolane 99%
2-Methyl-1,3-dithiolane
Purity: 99%
CAS No.: 5616-51-3
Molecular Formula: C4H8S2
Formula Weight: 120.23
Flash Point:
Formula:
C₄H₈S₂ Boiling Pt: 40 °C/2 mm Density: 1.15 Flash Pt: >100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01729666 CAS Number: 5616-51-3 EINECS: 227-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...