5,7-Dimethoxycoumarin 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Citropten, 5,7-Dimethoxy-2H-chromen-2-one

B23226-01 B23226-06 B23226-03
AAB23226-01EA 39.2 USD
AAB23226-01 AAB23226-06 AAB23226-03
5,7-Dimethoxycoumarin 98%
5,7-Dimethoxycoumarin

Purity: 98% CAS No.: 487-06-9 Molecular Formula: C11H10O4 Formula Weight: 206.20 Melting Point: 146-149° MDL No.: MFCD00006870

Formula: C₁₁H₁₀O₄
MW: 206.20 g/mol
Melting Pt: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00006870
CAS Number: 487-06-9
EINECS: 207-646-4
Merck Index: 14,5492

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR