2-Amino-4-(2-naphthyl)thiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Naphthalen-2-yl)thiazol-2-amine

B23223-03 B23223-06
AAB23223-03EA 26.8 USD
AAB23223-03 AAB23223-06
2-Amino-4-(2-naphthyl)thiazole 98%
2-Amino-4-(2-naphthyl)thiazole

Purity: 98% CAS No.: 21331-43-1 Molecular Formula: C13H10N2S Formula Weight: 226.30 Melting Point: 153-156° Beilstein Reference: 173533

Formula: C₁₃H₁₀N₂S
MW: 226.30 g/mol
Melting Pt: 153…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00046452
CAS Number: 21331-43-1
UN: 3077
ADR: "9",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR