4-(Trifluoromethylsulfonyl)benzonitrile 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22907-03
B22907-06
AAAB22907-03EA
45.2
USD
AAAB22907-03
AAAB22907-06
4-(Trifluoromethylsulfonyl)benzonitrile 97+%
4-(Trifluoromethylsulphonyl)benzonitrile
Purity: 97+%
CAS No.: 312-21-0
Molecular Formula: C8H4F3NO2S
Formula Weight: 235.18
Melting Point: 87-92°
UN No.: 3439
MDL No.: MFCD01631631
Formula:
C₈H₄F₃NO₂S Melting Pt: 87…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631631 CAS Number: 312-21-0 UN: 3439 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...