Citronellic acid 94%

Supplier: Thermo Scientific Chemicals
Warning

B22862-22 B22862-36 B22862-14
AAB22862-22EA 191 USD
AAB22862-22 AAB22862-36 AAB22862-14
Citronellic acid 94%
Citronellic acid

Purity: 94% CAS No.: 502-47-6 Molecular Formula: C10H18O2 Formula Weight: 170.25 Flash Point: 110°(230°F) Refractive Index: 1.4535 Boiling Point: 121-122°/1mm UN No.: 2810 MDL No.: MFCD00002728

Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
Boiling Pt: 121…122 °C/1 mm
Density: 0.923
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002728
CAS Number: 502-47-6
EINECS: 207-939-7
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR