3-(5-Methyl-2-furyl)butyraldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

B22671-06 B22671-14
AAB22671-22EA 405.12 USD
AAB22671-22 AAB22671-06 AAB22671-14
3-(5-Methyl-2-furyl)butyraldehyde 99%
3-(5-Methyl-2-furyl)butyraldehyde

Purity: 99% CAS No.: 31704-80-0 Molecular Formula: C9H12O2 Formula Weight: 152.19 Flash Point: 58°(136°F) Boiling Point: 93-94°/14mm UN No.: 1989 MDL No.: MFCD00209519

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 93…94 °C/14 mm
Density: 1.006
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00209519
CAS Number: 31704-80-0
EINECS: 250-771-4
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR