Ferulic acid methyl ester 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl 4-hydroxy-3-methoxycinnamate, Methyl 3-(4-hydroxy-3-methoxyphenyl)acrylate
B22657-04
B22657-09
AAB22657-04EA
54.4
USD
AAB22657-04
AAB22657-09
Ferulic acid methyl ester 99%
Ferulic acid methyl ester
Purity: 99%
CAS No.: 2309-07-1
Molecular Formula: C11H12O4
Formula Weight: 208.22
Melting Point: 62-65°
MDL No.: MFCD00017208
Formula:
C₁₁H₁₂O₄ MW: 208.22 g/mol Melting Pt: 62…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017208 CAS Number: 2309-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...