5-Methoxy-2-methylbenzoselenazole 97%

Supplier: Thermo Scientific Chemicals

B22411-03 B22411-06
AAB22411-03EA 32.6 USD
AAB22411-03 AAB22411-06
5-Methoxy-2-methylbenzoselenazole 97%
5-Methoxy-2-methylbenzoselenazole

Purity: 97% CAS No.: 2946-17-0 Molecular Formula: C9H9NOSe Formula Weight: 226.14 Flash Point: 110°(230°F) Refractive Index: 1.6390 Boiling Point: 115-117°/1mm UN No.: 3440 MDL No.: MFCD00005558

Formula: C₉H₉NOSe
Boiling Pt: 115…117 °C/1 mm
Density: 1.490
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005558
CAS Number: 2946-17-0
EINECS: 220-956-4
UN: 3440
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR