tert-Butyl methyl sulfide ≥98%

Supplier: Thermo Scientific Chemicals
Danger

B22390-06 B22390-14
AAB22390-06EA 43.5 USD
AAB22390-06 AAB22390-14
tert-Butyl methyl sulfide ≥98%
tert-Butyl methyl sulfide

Purity: 98+% CAS No.: 6163-64-0 Molecular Formula: C5H12S Formula Weight: 104.22 Flash Point: -3°(26°F) Refractive Index: 1.4400 Odor: Unpleasant Boiling Point: 101-102° UN No.: 1993 MDL No.: MFCD00008840

Formula: C₅H₁₂S
MW: 104.21 g/mol
Boiling Pt: 101…102 °C
Density: 0.826
Flash Pt: –3 °C (26 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008840
CAS Number: 6163-64-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR