5-Methyl-2-hepten-4-one predominantly trans 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22335-04
B22335-09
AAB22335-04EA
62.2
USD
AAB22335-04
AAB22335-09
5-Methyl-2-hepten-4-one predominantly trans 99%
5-Methyl-2-hepten-4-one
Purity: 99%
CAS No.: 81925-81-7
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 55°(131°F)
Refractive Index: 1.4440
Boiling Point: 169-171°
UN No.: 1224
MDL No.: MFCD00792509
Formula:
C₈H₁₄O MW: 126.20 g/mol Boiling Pt: 169…171 °C Flash Pt: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00792509 CAS Number: 81925-81-7 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...