Boc-Glu(OtBu)-OH 99%
Supplier: Thermo Scientific Chemicals
B22322-06
B22322-03
AAB22322-06EA
149
USD
AAB22322-06
AAB22322-03
Boc-Glu(OtBu)-OH 99%
Boc-Glu(OtBu)-OH
Purity: 99%
CAS No.: 13726-84-6
Molecular Formula: C14H25NO6
Formula Weight: 303.36
Melting Point: 110-112°
MDL No.: MFCD00038257
Formula:
C₁₄H₂₅NO₆ MW: 303.355 g/mol Melting Pt: 110…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038257 CAS Number: 13726-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...