3-Methyl-2-butenal 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 97%
CAS No.: 107-86-8
Molecular Formula: C5H8O
Formula Weight: 84.12
Storage Temperature: 2°C to 8°C
Flash Point: 37°(99°F)
Refractive Index: 1.4613
Odor: Characteristic
Melting Point: -20°
Boiling Point: 136°
Beilstein Reference: 1734740
UN No.: 2929
MDL No.: MFCD00010291
Formula:
C₅H₈O Boiling Pt: 136 °C Melting Pt: –20 °C Density: 0.88 Flash Pt: 37 °C(99 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00010291 CAS Number: 107-86-8 EINECS: 203-527-6 UN: 2920 ADR: "8",II Merck Index: 14,8448 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...