Cobalt(II) phosphate, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Warning

B22066-14 B22066-22
AAAB22066-14EA 61.3 USD
AAAB22066-14 AAAB22066-22
Cobalt(II) phosphate, anhydrous 98%
Cobalt(II) phosphate

Purity: 98% CAS No.: 13455-36-2 Molecular Formula: Co3O8P2 Formula Weight: 366.74 MDL No.: MFCD00016028

Formula: Co₃O₈P₂
MW: 366.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016028
CAS Number: 13455-36-2
EINECS: 236-655-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR