Dibismuth trisulphate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Bismuth (III) sulphate, Bismuth (III) sulfate

B22007-14 B22007-22
AAB22007-14EA 35.7 USD
AAB22007-14 AAB22007-22
Dibismuth trisulphate 99%
Dibismuth trisulphate

Purity: 99% CAS No.: 7787-68-0 Molecular Formula: Bi2O12S3 Formula Weight: 706.13 Melting Point: 405° dec. Merck Reference: 14,1286 MDL No.: MFCD00036140

Formula: Bi₂O₁₂S₃
Melting Pt: 405 °C (dec.)
Density: 5.08
Storage Temperature: Ambient
MDL Number: MFCD00036140
CAS Number: 7787-68-0
EINECS: 232-129-5
Merck Index: 14,1286

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR