1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Hydroxyphenyl)-5-mercaptotetrazole

B22003-06 B22003-22 B22003-14
AAB22003-06EA 29.8 USD
AAB22003-06 AAB22003-22 AAB22003-14
1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole 96%
1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole

Purity: 96% CAS No.: 52431-78-4 Molecular Formula: C7H6N4OS Formula Weight: 194.21 Melting Point: 169-171° MDL No.: MFCD00132898

Formula: C₇H₆N₄OS
MW: 194.21 g/mol
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00132898
CAS Number: 52431-78-4
EINECS: 257-909-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR