N-Boc-D-glutamic acid 1-benzyl ester 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Benzyl N-Boc-D-glutamate, Boc-D-Glu-OBzl, N-Boc-D-glutamic Acid 1-Benzyl Ester, N-(tert-Butoxycarbonyl)-D-glutamic Acid 1-Benzyl Ester

B21944-03 B21944-06
AAB21944-03EA 35.6 USD
AAB21944-03 AAB21944-06
N-Boc-D-glutamic acid 1-benzyl ester 99%
N-Boc-D-glutamic acid 1-benzyl ester

Purity: 99% CAS No.: 34404-30-3 Molecular Formula: C17H23NO6 Formula Weight: 337.37 MDL No.: MFCD00038266

Formula: C₁₇H₂₃NO₆
MW: 337.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038266
CAS Number: 34404-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR