Tolbutamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-(Butylcarbamoyl)-4-methylbenzenesulphonamide
B21698-18
B21698-30
B21698-09
AAB21698-18EA
182
USD
AAB21698-18
AAB21698-30
AAB21698-09
Tolbutamide 98%
Tolbutamide
A first-generation potassium channel blocker
Purity: 98%
CAS No.: 64-77-7
Molecular Formula: C12H18N2O3S
Formula Weight: 270.35
Melting Point: 128-130°
Beilstein Reference: 1984428
Merck Reference: 14,9507
MDL No.: MFCD00027169
Formula:
C₁₂H₁₈N₂O₃S MW: 270.35 Melting Pt: 128…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00027169 CAS Number: 64-77-7 EINECS: 200-594-3 Merck Index: 14,9507 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...