2-Thioxanthine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Thioxo-2,3-dihydro-1H-purin-6(7H)-one
B21606-03
B21606-06
AAB21606-03EA
114
USD
AAB21606-03
AAB21606-06
2-Thioxanthine 98+%
2-Thioxanthine
Purity: 98+%
CAS No.: 2487-40-3
Molecular Formula: C5H4N4OS
Formula Weight: 168.17
Melting Point:
Formula:
C₅H₄N₄OS MW: 168.17 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031505 CAS Number: 2487-40-3 EINECS: 219-636-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...