Cyclohexyl hexanoate 99%

Supplier: Thermo Scientific Chemicals

B21547-22 B21547-14
AAB21547-22EA 138 USD
AAB21547-22 AAB21547-14
Cyclohexyl hexanoate 99%
Cyclohexyl hexanoate

Purity: 99% CAS No.: 6243-10-3 Molecular Formula: C12H22O2 Formula Weight: 198.31 MDL No.: MFCD00046356

Formula: C₁₂H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00046356
CAS Number: 6243-10-3
EINECS: 228-354-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR