Glibenclamide 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
5-Chloro-N-(4-(N-(cyclohexylcarbamoyl)sulphamoyl)phenethyl)-2-methoxybenzamide, 5-Chloro-N-[4-(3-cyclohexylureidosulphonyl)phenethyl]-2-methoxybenzamide, Glyburide
B21459-14
B21459-22
AAB21459-14EA
97.46
USD
AAB21459-14
AAB21459-22
Glibenclamide 99%
Glibenclamide
ATP-dependent potassium channel (KATP) and cystic fibrosis transmembrane conductance regulator (CFTR) chloride channel blocker
Purity: 99%
CAS No.: 10238-21-8
Molecular Formula: C23H28ClN3O5S
Formula Weight: 494.01
Storage Temperature: 2°C to 8°C
Melting Point: 171-174°
MDL No.: MFCD00056625
Formula:
C₂₃H₂₈ClN₃O₅S MW: 494.00 g/mol Melting Pt: 171…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00056625 CAS Number: 10238-21-8 EINECS: 233-570-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...