N-Vinyl-ε-caprolactam 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Vinylazepan-2-one

B21311-09 B21311-18
AAB21311-09EA 33.5 USD
AAB21311-09 AAB21311-18
N-Vinyl-ε-caprolactam 99%
N-Vinyl-epsilon-caprolactam

Purity: 99% CAS No.: 2235-00-9 Molecular Formula: C8H13NO Formula Weight: 139.20 Flash Point: 101°(214°F) Melting Point: 32-36° Boiling Point: 128°/21mm MDL No.: MFCD00080693

Formula: C₈H₁₃NO
MW: 139.20 g/mol
Boiling Pt: 128 °C/21 mm
Melting Pt: 32…36 °C
Density: 1.029
Flash Pt: 101 °C(214 °F)
Storage Temperature: Ambient
MDL Number: MFCD00080693
CAS Number: 2235-00-9
EINECS: 218-787-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR