1-(Trimethylsilyl)-1,2,4-triazole 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21105-14
B21105-06
AAB21105-14EA
287
USD
AAB21105-14
AAB21105-06
1-(Trimethylsilyl)-1,2,4-triazole 95%
1-(Trimethylsilyl)-1,2,4-triazole
Purity: 95%
CAS No.: 18293-54-4
Molecular Formula: C5H11N3Si
Formula Weight: 141.25
Flash Point: -5°(23°F)
Refractive Index: 1.4610
Boiling Point: 73-75°/12mm
UN No.: 1993
MDL No.: MFCD00046070
Formula:
C₅H₁₁N₃Si Boiling Pt: 73…75 °C/12 mm Density: 0.98 Flash Pt: -5 °C(23 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00046070 CAS Number: 18293-54-4 EINECS: 242-170-0 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...