2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Van Boom's Reagent, Salicylchlorophosphite

B21086-14 B21086-06
AAB21086-14EA 262 USD
AAB21086-14 AAB21086-06
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one 97%
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one

Purity: 97% CAS No.: 5381-99-7 Molecular Formula: C7H4ClO3P Formula Weight: 202.54 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 36-40° Boiling Point: 127-128°/11mm UN No.: 3261 MDL No.: MFCD00013353

Formula: C₇H₄ClO₃P
Boiling Pt: 127…128 °C/11 mm
Melting Pt: 36…40 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013353
CAS Number: 5381-99-7
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR