6-Hydroxy-2-methylpyridazin-3(2H)-one 97%
Supplier: Thermo Scientific Chemicals
B20975-14
B20975-06
AAB20975-14EA
332
USD
AAB20975-14
AAB20975-06
6-Hydroxy-2-methylpyridazin-3(2H)-one 97%
6-Hydroxy-2-methylpyridazin-3(2H)-one
Purity: 97%
CAS No.: 5436-01-1
Molecular Formula: C5H6N2O2
Formula Weight: 126.11
Melting Point: 216-219°
MDL No.: MFCD00234108
Formula:
C₅H₆N₂O₂ Melting Pt: 216…219 °C Storage Temperature: Ambient |
MDL Number:
MFCD00176268 CAS Number: 5436-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...