2-(4-Methylphenyl)ethanol 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-(p-Tolyl)ethanol
B20918-14
B20918-03
B20918-06
AAB20918-14EA
504
USD
AAB20918-14
AAB20918-03
AAB20918-06
2-(4-Methylphenyl)ethanol 99%
2-(4-Methylphenyl)ethanol
Purity: 99%
CAS No.: 699-02-5
Molecular Formula: C9H12O
Formula Weight: 136.20
Flash Point: 107°(225°F)
Refractive Index: 1.5260
Boiling Point: 244-245°
MDL No.: MFCD00002904
Formula:
C₉H₁₂O MW: 136.20 g/mol Boiling Pt: 244…245 °C Density: 0.980 Flash Pt: 107 °C(225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002904 CAS Number: 699-02-5 EINECS: 211-824-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...