Di-n-propyl succinate 98%

Supplier: Thermo Scientific Chemicals
Warning

B20441-09 B20441-18
AAAB20441-09EA 39.9 USD
AAAB20441-09 AAAB20441-18
Di-n-propyl succinate 98%
Di-n-propyl succinate

Purity: 98% CAS No.: 925-15-5 Molecular Formula: C10H18O4 Formula Weight: 202.25 Odor: Pleasant Boiling Point: 250-252° MDL No.: MFCD00015213

Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 250…252 °C
Density: 1.00
Storage Temperature: Ambient
MDL Number: MFCD00015213
CAS Number: 925-15-5
EINECS: 213-114-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR