[(4-Fluorophenyl)sulfonyl]acetonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19916-06
A19916-03
AAA19916-06EA
250
USD
AAA19916-06
AAA19916-03
AAA19916-14
[(4-Fluorophenyl)sulfonyl]acetonitrile 97%
[(4-Fluorophenyl)sulfonyl]acetonitrile
Purity: 97%
CAS No.: 32083-66-2
Molecular Formula: C8H6FNO2S
Formula Weight: 199.20
Melting Point: 89-93°
UN No.: 3439
MDL No.: MFCD00179339
Formula:
C₈H₆FNO₂S Melting Pt: 89…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00179339 CAS Number: 32083-66-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...