Geranyl acetate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-yl acetate

A19864-14 A19864-22
AAA19864-14EA 43.14 USD
AAA19864-14 AAA19864-22
Geranyl acetate 98%
Geranyl acetate

Purity: 98% CAS No.: 105-87-3 Molecular Formula: (CH3)2C=CHCH2CH2C(CH3)=CHCH2O2CCH3 Formula Weight: 196.29 Storage Temperature: 2°C to 8°C Flash Point: 104°(219°F) Physical Form: Liquid Density: 0.91 Refractive Index: 1.461 Appearance: Colorless Odor: Pleasant Boiling Point: 137-139°/25mm MDL No.: MFCD00015037

Formula: C₁₂H₂₀O₂
MW: 196.29 g/mol
Boiling Pt: 137…139 °C/25 mm
Density: 0.91
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015037
CAS Number: 105-87-3
EINECS: 203-341-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR