4-Butylbenzamide 97%

Supplier: Thermo Scientific Chemicals

A19516-03 A19516-06
AAA19516-14EA 346.29 USD
AAA19516-14 AAA19516-03 AAA19516-06
4-Butylbenzamide 97%
4-Butylbenzamide

Purity: 97% CAS No.: 107377-07-1 Molecular Formula: C11H15NO Formula Weight: 177.25 Melting Point: 127-129° MDL No.: MFCD00221466

Formula: C₁₁H₁₅NO
Melting Pt: 127…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00221466
CAS Number: 107377-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR