DL-Malic acid monosodium salt 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium hydrogen DL-malate
A19428-18
A19428-30
AAAA19428-18EA
58.1
USD
AAAA19428-18
AAAA19428-30
DL-Malic acid monosodium salt 98%
DL-Malic acid monosodium salt
Purity: 98%
CAS No.: 57467-17-1
Molecular Formula: C4H5NaO5
Formula Weight: 156.07
Melting Point: 210° dec.
MDL No.: MFCD00798569
Formula:
C₄H₅NaO₅ MW: 156.07 g/mol Melting Pt: 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00798569 CAS Number: 57467-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...