(4-tert-Butylphenoxy)acetyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-(4-(tert-Butyl)phenoxy)acetyl chloride
A19408-09
A19408-18
AAA19408-09EA
54.2
USD
AAA19408-09
AAA19408-18
(4-tert-Butylphenoxy)acetyl chloride 98%
(4-tert-Butylphenoxy)acetyl chloride
Purity: 98%
CAS No.: 90734-55-7
Molecular Formula: C12H15O2Cl
Formula Weight: 226.71
Refractive Index: 1.5165
Boiling Point: 130-132°/5mm
UN No.: 3265
MDL No.: MFCD00798568
Formula:
C₁₂H₁₅ClO₂ MW: 226.71 g/mol Boiling Pt: 130…132 °C/5 mm Storage Temperature: Ambient |
MDL Number:
MFCD00798568 CAS Number: 90734-55-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...