cis-3-Hexenyl acetate 99%

Supplier: Thermo Scientific Chemicals
Warning

A19380-22 A19380-14
AAA19380-22EA 149.19 USD
AAA19380-22 AAA19380-14
cis-3-Hexenyl acetate 99%
cis-3-Hexenyl acetate

Purity: 99% CAS No.: 3681-71-8 Molecular Formula: C8H14O2 Formula Weight: 142.20 Flash Point: 59°(138°F) Refractive Index: 1.4270 Boiling Point: 169° Beilstein Reference: 1721854 UN No.: 3272 MDL No.: MFCD00036563

Formula: C₈H₁₄O₂
MW: 142.20 g/mol
Boiling Pt: 169 °C
Density: 0.899
Flash Pt: 59 °C(138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036563
CAS Number: 3681-71-8
EINECS: 222-960-1
UN: 3272
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR