Neryl acetate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(Z)-3,7-Dimethylocta-2,6-dien-1-yl acetate
A19282-22
A19282-14
AAA19282-22EA
116
USD
AAA19282-22
AAA19282-14
Neryl acetate 98%
Neryl acetate
Purity: 98%
CAS No.: 141-12-8
Molecular Formula: C12H20O2
Formula Weight: 196.29
Storage Temperature: 2°C to 8°C
Flash Point: 98°(208°F)
Refractive Index: 1.4600
Odor: Pleasant
Boiling Point: 133-135°/25mm
Beilstein Reference: 1722814
MDL No.: MFCD00063205
Formula:
C₁₂H₂₀O₂ Boiling Pt: 133…135 °C/25 mm Density: 0.910 Flash Pt: 98 °C(208 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00063205 CAS Number: 141-12-8 EINECS: 205-459-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...