3-Hepten-2-one 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hept-3-en-2-one
A19086-09
A19086-18
AAA19086-09EA
62
USD
AAA19086-09
AAA19086-18
3-Hepten-2-one 97%
3-Hepten-2-one
Purity: 97%
CAS No.: 1119-44-4
Molecular Formula: C7H12O
Formula Weight: 112.16
Storage Temperature: 2°C to 8°C
Flash Point: 49°(120°F)
Refractive Index: 1.4450
Boiling Point: 156°
Beilstein Reference: 1739239
UN No.: 1224
MDL No.: MFCD00015564
Formula:
C₇H₁₂O MW: 112.17 g/mol Boiling Pt: 156 °C Density: 0.860 Flash Pt: 49 °C (120 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00015564 CAS Number: 1119-44-4 EINECS: 214-278-8 UN: 1224 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...