Trithiocyanuric acid 95%

Supplier: Thermo Scientific Chemicals
Warning

A19080-14 A19080-06 A19080-22
AAA19080-14EA 146 USD
AAA19080-14 AAA19080-06 AAA19080-22
Trithiocyanuric acid 95%
Trithiocyanuric acid

Purity: 95% CAS No.: 638-16-4 Molecular Formula: C3H3N3S3 Formula Weight: 177.27 Melting Point: 300° MDL No.: MFCD00006052

Formula: C₃H₃N₃S₃
MW: 177.27 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006052
CAS Number: 638-16-4
EINECS: 211-322-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR