4,4'-Methylenebis(2,6-di-tert-butylphenol) 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Bis(3,5-di-tert-butyl-4-hydroxyphenyl)methane
A18910-22
A18910-18
AAA18910-22EA
47.8
USD
AAA18910-22
AAAA18910-18
4,4'-Methylenebis(2,6-di-tert-butylphenol) 98%
4,4'-Methylenebis(2,6-di-tert-butylphenol)
Fieser: 4,337
Purity: 98%
CAS No.: 118-82-1
Molecular Formula: C29H44O2
Formula Weight: 424.67
Melting Point: 154-158°
Boiling Point: 289°/40mm
Beilstein Reference: 1916919
MDL No.: MFCD00008822
Formula:
C₂₉H₄₄O₂ MW: 424.67 g/mol Boiling Pt: 289 °C/40 mm Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008822 CAS Number: 118-82-1 EINECS: 204-279-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...