2,4,5,6-Tetraaminopyrimidine sulfate 97%

Supplier: Thermo Scientific Chemicals
Warning

A18788-18 A18788-09 A18788-30
AAA18788-18EA 84.4 USD
AAA18788-18 AAA18788-09 AAA18788-30
2,4,5,6-Tetraaminopyrimidine sulfate 97%
2,4,5,6-Tetraaminopyrimidine sulphate

Used as an intermediate in the production of Methotrexate.


Purity: 97% CAS No.: 5392-28-9 Molecular Formula: C4H8N6.H2SO4 Formula Weight: 238.23 Melting Point: 300° Beilstein Reference: 3785189

Formula: C₄H₁₀N₆O₄S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012786
CAS Number: 5392-28-9
EINECS: 226-393-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR