Geranic acid (sum of isomers) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18750-22
A18750-14
AAA18750-22EA
202.94
USD
AAA18750-22
AAA18750-14
Geranic acid (sum of isomers) 98%
Geranic acid (sum of isomers)
Purity: 90+%
CAS No.: 459-80-3
Molecular Formula: C10H16O2
Formula Weight: 168.24
Flash Point: 133°(271°F)
Refractive Index: 1.4840
Boiling Point: 249-251°
Beilstein Reference: 1763804
MDL No.: MFCD00036493
Formula:
C₁₀H₁₆O₂ Boiling Pt: 249…251 °C Density: 0.955 Flash Pt: 133 °C(271 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00036493 CAS Number: 459-80-3 EINECS: 207-299-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...