2-Phenylquinoline-4-carboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Phenyl-4-quinolinecarboxylic acid, Cinchophen

A18734-22 A18734-14
AAA18734-22EA 160 USD
AAA18734-22 AAA18734-14
2-Phenylquinoline-4-carboxylic acid 99%
2-Phenylquinoline-4-carboxylic acid

Insoluble in water


Purity: 99% CAS No.: 132-60-5 Molecular Formula: C16H11NO2 Formula Weight: 249.27 Odor: Odorless Melting Point: 212-218° Beilstein Reference: 192803 Merck Reference: 14,2288 MDL No.: MFCD00006750

Formula: C₁₆H₁₁NO₂
MW: 249.27 g/mol
Melting Pt: 212…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00006750
CAS Number: 132-60-5
EINECS: 205-067-1
Merck Index: 14,2288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR