4-Methoxytrityl alcohol 94%

Supplier: Thermo Scientific Chemicals

Synonyms: (4-Methoxyphenyl)diphenylmethanol

A18620-30 A18620-18 A18620-09
AAA18620-30EA 724 USD
AAA18620-30 AAA18620-18 AAA18620-09
4-Methoxytrityl alcohol 94%
4-Methoxytrityl alcohol

Purity: 94% CAS No.: 847-83-6 Molecular Formula: C20H18O2 Formula Weight: 290.36 MDL No.: MFCD00087962

Formula: C₂₀H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00087962
CAS Number: 847-83-6
EINECS: 212-693-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR