Phenyl trans-styryl sulfone 98%

Supplier: Thermo Scientific Chemicals

A18556-06 A18556-03
AAA18556-06EA 232 USD
AAA18556-06 AAA18556-03
Phenyl trans-styryl sulfone 98%
Phenyl trans-styryl sulfone

Purity: 98% CAS No.: 16212-06-9 Molecular Formula: C14H12O2S Formula Weight: 244.32 Melting Point: 74-76° MDL No.: MFCD00159177

Formula: C₁₄H₁₂O₂S
MW: 244.31 g/mol
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00159177
CAS Number: 16212-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR