2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-4-hydroxy-6-trifluoromethylpyrimidine

A18533-06 A18533-03
AAA18533-06EA 196 USD
AAA18533-06 AAA18533-03
2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine 97%
2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine

Purity: 97% CAS No.: 1513-69-5 Molecular Formula: C5H4F3N3O Formula Weight: 179.10 Melting Point: 288-292° Beilstein Reference: 3608100 MDL No.: MFCD00068093

Formula: C₅H₄F₃N₃O
MW: 179.1 g/mol
Melting Pt: 288…292 °C
Storage Temperature: Ambient
MDL Number: MFCD00068093
CAS Number: 1513-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR