Zinc chromate

Supplier: Thermo Scientific Chemicals
Danger

A18178-30 A18178-0B
AAAA18178-30EA 89.6 USD
AAAA18178-30 AAAA18178-0B
Zinc chromate
Zinc chromate

CAS No.: 13530-65-9 Molecular Formula: ZnCrO4 Formula Weight: 181.37 UN No.: 3077 MDL No.: MFCD00036281

Formula: ZnCrO₄
Density: 3.4
Storage Temperature: Ambient
MDL Number: MFCD00036281
CAS Number: 13530-65-9
EINECS: 236-878-9
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR