2-Chlorothioxanthone 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Chlorothioxanthen-9-one, 2-Chloro-9H-thioxanthen-9-one
A18131-14
A18131-22
A18131-06
AAA18131-14EA
64.4
USD
AAA18131-14
AAA18131-22
AAA18131-06
2-Chlorothioxanthone 99%
2-Chlorothioxanthone
Purity: 99%
CAS No.: 86-39-5
Molecular Formula: C13H7ClOS
Formula Weight: 246.72
Flash Point: 91°(196°F)
Melting Point: 151-154°
MDL No.: MFCD00005067
Formula:
C₁₃H₇ClOS MW: 246.72 g/mol Melting Pt: 151…154 °C Flash Pt: 91 °C(196 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005067 CAS Number: 86-39-5 EINECS: 201-667-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...