N-Methyl-ε-caprolactam 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Methyl-2-azacycloheptanone, 1-Methylazepan-2-one

A17776-22 A17776-14
AAA17776-22EA 148 USD
AAA17776-22 AAA17776-14
N-Methyl-ε-caprolactam 96%
N-Methyl-epsilon-caprolactam

Purity: 96% CAS No.: 2556-73-2 Molecular Formula: C7H13NO Formula Weight: 127.18 Flash Point: 103°(217°F) Refractive Index: 1.4840 Boiling Point: 106-108°/6mm MDL No.: MFCD00003263

Formula: C₇H₁₃NO
MW: 127.19 g/mol
Boiling Pt: 106…108 °C/6 mm
Density: 0.991
Flash Pt: 103 °C(217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003263
CAS Number: 2556-73-2
EINECS: 219-874-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR