8-Hydroxyjulolidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2,3,5,6,7-Hexahydropyrido[3,2,1-ij]quinolin-8-ol, 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol

A17692-06 A17692-03
AAA17692-06EA 225 USD
AAA17692-06 AAA17692-03
8-Hydroxyjulolidine 97%
8-Hydroxyjulolidine

Purity: 97% CAS No.: 41175-50-2 Molecular Formula: C12H15NO Formula Weight: 189.26 Melting Point: 132-135° Beilstein Reference: 1530423 MDL No.: MFCD00006918

Formula: C₁₂H₁₅NO
MW: 189.26 g/mol
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00006918
CAS Number: 41175-50-2
EINECS: 255-247-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR