2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid-N-hydroxysuccinimide ester ≥98%
Supplier: Thermo Scientific Chemicals
A17667-01
A17667-03
AAA17667-01EA
95.7
USD
AAA17667-01
AAAA17667-03
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid-N-hydroxysuccinimide ester ≥98%
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid-N-hydroxysuccinimide ester
Purity: 98+%
CAS No.: 37558-29-5
Molecular Formula: C13H17N2O5
Formula Weight: 281.29
Physical Form: Crystalline powder
Appearance: Yellow
Melting Point: 173-177°
MDL No.: MFCD00066369
Formula:
C₁₃H₁₇N₂O₅ MW: 281.29 g/mol Melting Pt: 173…177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066369 CAS Number: 37558-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...