Nonenylsuccinic anhydride (mixture of branched chain isomers) 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: NSA, Isononenylsuccinic anhydride

A17607-30 A17607-18
AAA17607-30EA 172 USD
AAA17607-30 AAA17607-18
Nonenylsuccinic anhydride (mixture of branched chain isomers) 96%
Nonenylsuccinic anhydride (mixture of branched chain isomers)

Purity: 96% CAS No.: 28928-97-4 Molecular Formula: C13H20O3 Formula Weight: 224.30 Flash Point: 146°(295°F) Boiling Point: 126-130°/0.15mm MDL No.: MFCD00014552

Formula: C₁₃H₂₀O₃
MW: 224.30 g/mol
Boiling Pt: 126…130 °C/0.15 mm
Density: 1.032
Flash Pt: 146 °C(295 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014552
CAS Number: 28928-97-4
EINECS: 249-317-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR