Ammonium magnesium phosphate hexahydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: AMMONIUM MAGNESIUM PHOSPHATE HEXAHYDRATE

A17497-22 A17497-36
AAAA17497-22EA 93.55 USD
AAAA17497-22 AAAA17497-36
Ammonium magnesium phosphate hexahydrate 98%
Ammonium magnesium phosphate hexahydrate

Purity: 98% CAS No.: 13478-16-5 Molecular Formula: NH4MgPO4.6H2O Formula Weight: 245.41 (137.32anhy) MDL No.: MFCD00150461

Formula: NH₄MgPO₄·6H₂O
MW: 245.41 g/mol (137.32 anhy)
Density: 1.711
Storage Temperature: Ambient
MDL Number: MFCD00150461
CAS Number: 13478-16-5
EINECS: 232-075-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR