Janus green B
Supplier: Thermo Scientific Chemicals
A17391-06
A17391-14
AAA17391-06EA
66.65
USD
AAA17391-06
AAA17391-14
Janus green B
Janus green B
CAS No.: 2869-83-2
Molecular Formula: C30H31ClN6
Formula Weight: 511.07
MDL No.: MFCD00011758
Formula:
C₃₀H₃₁ClN₆ MW: 511.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011758 CAS Number: 2869-83-2 EINECS: 220-695-6 Merck Index: 14,5255 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...